accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOOOF

CAS RN 652-12-0

IUPAC Name: tetrafluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: tetrafluorophthalic anhydride
Molecular Weight (g/mol): 220.08
Molecular Formula: C8F4O3
InChi Key: BJDDKZDZTHIIJB-UHFFFAOYSA-N
SMILES: FC1=C2C(=O)OC(=O)C2=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)