Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluorophthalic Anhydride 95.0+%, TCI America™
SDP

Catalog No. T23815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23815G 5 g
T23811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23815G Supplier TCI America Supplier No. T23815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 652-12-0
Molecular Formula C8F4O3
Molecular Weight (g/mol) 220.079
MDL Number MFCD00039697
InChI Key BJDDKZDZTHIIJB-UHFFFAOYSA-N
Synonym tetrafluorophthalic anhydride, 3,4,5,6-tetrafluorophthalic anhydride, 4,5,6,7-tetrafluoroisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4,5,6,7-tetrafluoro, tetrafluorophthalicanhydride, pubchem1951, acmc-209nps, tetrafluoroisobenzofuran-1,3-dione, 3,4,5,6-tetrafluorophthalicanhydride
PubChem CID 69545
IUPAC Name 4,5,6,7-tetrafluoro-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=O)OC2=O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 220.08
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorophthalic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.