accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 652-39-1

IUPAC Name: 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-fluoro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 166.11
Molecular Formula: C8H3FO3
InChi Key: WWJAZKZLSDRAIV-UHFFFAOYSA-N
SMILES: FC1=CC=CC2=C1C(=O)OC2=O
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)