accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 652145-29-4

IUPAC Name: 2-cyano-3-{4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,4H,8bH-cyclopenta[b]indol-7-yl}prop-2-enoic acid
Synonyms: 2-cyano-3-{4-[4-(2,2-diphenylethenyl)phenyl]-1H,2H,3H,3aH,8bH-cyclopenta[b]indol-7-yl}prop-2-enoic acid
Molecular Weight (g/mol): 508.62
Molecular Formula: C35H28N2O2
InChi Key: GOTRYMLNXIJMCB-UHFFFAOYNA-N
SMILES: OC(=O)C(=CC1=CC2=C(C=C1)N(C1CCCC21)C1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results