accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNN

CAS RN 6523-49-5

IUPAC Name: 4-(1H-1,2,4-triazol-1-yl)aniline
Synonyms: 4-(1,2,4-triazol-1-yl)aniline
Molecular Weight (g/mol): 160.18
Molecular Formula: C8H8N4
InChi Key: BLNPEJXSNSBBNM-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1C=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)