Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-1,2,4-triazol-1-yl)aniline, 97%, Thermo Scientific™

Catalog No. CC18614EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18614EA 10 g
CC18614DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC18614EA Supplier Thermo Scientific Chemicals Supplier No. CC18614EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6523-49-5
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00171470
InChI Key BLNPEJXSNSBBNM-UHFFFAOYSA-N
Synonym 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
PubChem CID 735826
IUPAC Name 4-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC=C1N)N2C=NC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.