accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65253-04-5

IUPAC Name: (1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
Synonyms: (1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
Molecular Weight (g/mol): 232.37
Molecular Formula: C16H24O
InChi Key: WTQIZFCJMGWUGZ-BPLDGKMQSA-N
SMILES: C[C@@H]1CC[C@H]([C@H](O)C1)C(C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)