Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-8-Phenylmenthol 98%, Thermo Scientific™

Catalog No. AC294872500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294872500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294872500 Supplier Thermo Scientific Chemicals Supplier No. 294872500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65253-04-5
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.367
InChI Key WTQIZFCJMGWUGZ-BPLDGKMQSA-N
Synonym --8-phenylmenthol, --phenmenthol, 1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexanol, cyclohexanol, 5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r, 1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexan-1-ol, 1r,2s,5r-5-methyl-2-1-methyl-1-phenylethyl cyclohexanol, cyclohexanol,5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r
PubChem CID 2725001
IUPAC Name (1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)(C)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.5295 to 1.5315 (20°C, 589nm)
Quantity 250 mg
Fieser 11,412; 12,389; 13,243; 14,259; 15,260; 16,276; 17,282
Specific Rotation -25° to -27° (20°C, 589nm) (c=2, C2H5OH)
Formula Weight 232.37
Percent Purity ≥97.5%
Physical Form Oily Liquid
Chemical Name or Material (-)-8-Phenylmenthol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.