Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-8-Phenylmenthol 98.0+%, TCI America™
SDP

Catalog No. P2327200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2327200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2327200MG Supplier TCI America Supplier No. P2327200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65253-04-5
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.367
MDL Number MFCD00010501
InChI Key WTQIZFCJMGWUGZ-BPLDGKMQSA-N
Synonym --8-phenylmenthol, --phenmenthol, 1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexanol, cyclohexanol, 5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r, 1r,2s,5r-5-methyl-2-2-phenylpropan-2-yl cyclohexan-1-ol, 1r,2s,5r-5-methyl-2-1-methyl-1-phenylethyl cyclohexanol, cyclohexanol,5-methyl-2-1-methyl-1-phenylethyl-, 1r,2s,5r
PubChem CID 2725001
IUPAC Name (1R,2S,5R)-5-methyl-2-(2-phenylpropan-2-yl)cyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)(C)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 85°C
Quantity 200 mg
Formula Weight 232.37
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-8-Phenylmenthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.