accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONON

CAS RN 6574-15-8

IUPAC Name: 1-(4-nitrophenyl)piperidine
Synonyms: N-(P-nitrophenyl)piperidine
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: SGPLAXFUDTWHRS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results