accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CN

CAS RN 6575-13-9

IUPAC Name: 2,6-dimethylbenzonitrile
Synonyms: 2,6-dimethylbenzonitrile
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: QSACPWSIIRFHHR-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1C#N
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)