Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylbenzonitrile, 99%, Thermo Scientific™

Catalog No. AC355590250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC355590250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC355590250 Supplier Thermo Scientific Chemicals Supplier No. 355590250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6575-13-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
InChI Key QSACPWSIIRFHHR-UHFFFAOYSA-N
Synonym benzonitrile, 2,6-dimethyl, unii-1zh37s5b0z, 2,6-dimethylbenzenecarbonitrile, 2,6-dimethyl-benzonitrile, 2-cyano-m-xylene, ksc494c5t, 2,6-dimethylbenz0nitrile, 3-cyano-2,4-dimethylphenyl
PubChem CID 81057
IUPAC Name 2,6-dimethylbenzonitrile
SMILES CC1=C(C(=CC=C1)C)C#N

Specifications

Melting Point 88.0°C to 92.0°C
Quantity 25 g
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylbenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 229-503-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.