accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 65954-42-9

IUPAC Name: 1-(pyren-1-yl)ethan-1-ol
Synonyms: 1-(pyren-1-yl)ethanol
Molecular Weight (g/mol): 246.31
Molecular Formula: C18H14O
InChi Key: LFCWFVPSUMBNQM-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results