accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOOClClCl

CAS RN 66065-85-8

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2,2,2-trichloroethyl carbonate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2,2,2-trichloroethyl carbonate
Molecular Weight (g/mol): 290.48
Molecular Formula: C7H6Cl3NO5
InChi Key: WBZXNGAFYBGQFE-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)COC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)