accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNSONH2OFFFOCH3

CAS RN 660868-91-7

IUPAC Name: 5-(5,6-dimethoxy-1H-1,3-benzodiazol-1-yl)-3-{[2-(trifluoromethyl)phenyl]methoxy}thiophene-2-carboxamide
Synonyms: 5-(5,6-dimethoxy-1,3-benzodiazol-1-yl)-3-{[2-(trifluoromethyl)phenyl]methoxy}thiophene-2-carboxamide
Molecular Weight (g/mol): 477.46
Molecular Formula: C22H18F3N3O4S
InChi Key: JSKUWFIZUALZLX-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2N(C=NC2=C1)C1=CC(OCC2=CC=CC=C2C(F)(F)F)=C(S1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)