accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6610-29-3

IUPAC Name: 3-amino-1-methylthiourea
Synonyms: 4-methyl-3-thiosemicarbazide
Molecular Weight (g/mol): 105.16
Molecular Formula: C2H7N3S
InChi Key: PTVZQOAHCSKAAS-UHFFFAOYSA-N
SMILES: CNC(=S)NN
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)