accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6625-46-3

IUPAC Name: disodium 5-acetamido-3-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium 5-acetamido-3-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molecular Weight (g/mol): 539.44
Molecular Formula: C19H15N3Na2O9S2
InChi Key: JIAWPRMYSNTTFB-UHFFFAOYSA-L
SMILES: [Na+].[Na+].COC1=CC=CC=C1NN=C1C(=O)C2=C(NC(C)=O)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results