accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6628-04-2

IUPAC Name: 2-methylquinolin-4-amine
Synonyms: 2-methyl-quinolin-4-ylamine
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: COCFIBRMFPWUDW-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C2C=CC=CC2=N1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)