Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-methylquinoline 97.0+%, TCI America™
SDP

Catalog No. A04155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04155G 5 g
A041525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04155G Supplier TCI America Supplier No. A04155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6628-04-2 | C10H10N2 | 158.20 g/mol

Chemical Identifiers

CAS 6628-04-2
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00006759
InChI Key COCFIBRMFPWUDW-UHFFFAOYSA-N
Synonym 4-aminoquinaldine, 4-amino-2-methylquinoline, 2-methyl-4-aminoquinoline, 4-quinolinamine, 2-methyl, 2-methyl-4-quinolylamine, 2-methyl-quinolin-4-ylamine, 4-aminoquinaldine 4, 2-methyl-4-quinolinamine, acmc-1b7ou
PubChem CID 81116
IUPAC Name 2-methylquinolin-4-amine
SMILES CC1=NC2=CC=CC=C2C(=C1)N

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methylquinoline
EINECSNumber (5)-3738
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.