Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoquinaldine 98%, Thermo Scientific™

Catalog No. AC104590250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC104590250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC104590250 Supplier Thermo Scientific Chemicals Supplier No. 104590250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6628-04-2
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
InChI Key COCFIBRMFPWUDW-UHFFFAOYSA-N
Synonym 4-aminoquinaldine, 4-amino-2-methylquinoline, 2-methyl-4-aminoquinoline, 4-quinolinamine, 2-methyl, 2-methyl-4-quinolylamine, 2-methyl-quinolin-4-ylamine, 4-aminoquinaldine 4, 2-methyl-4-quinolinamine, acmc-1b7ou
PubChem CID 81116
IUPAC Name 2-methylquinolin-4-amine
SMILES CC1=NC2=CC=CC=C2C(=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 165°C to 169°C
Color Beige to Yellow
Infrared Spectrum Authentic
Quantity 25 g
Beilstein 22, 453
Formula Weight 158.2
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 4-Aminoquinaldine, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.