accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHH2NHOOOHO

CAS RN 6629-60-3

IUPAC Name: ethylhydrazine; oxalic acid
Synonyms: ethylhydrazine; oxalic acid
Molecular Weight (g/mol): 150.13
Molecular Formula: C4H10N2O4
InChi Key: DUMHBFMURBWDPC-UHFFFAOYSA-N
SMILES: CCNN.OC(=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)