accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6630-01-9

IUPAC Name: 1-tert-butyl-3-ethyl-5-methylbenzene
Synonyms: 1-tert-butyl-3-ethyl-5-methylbenzene
Molecular Weight (g/mol): 176.3
Molecular Formula: C13H20
InChi Key: URDKTSKLGNWKDB-UHFFFAOYSA-N
SMILES: CCC1=CC(C)=CC(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results