accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 666-40-0

IUPAC Name: 2,3-dibromo-1,1,3,3-tetrafluoroprop-1-ene
Synonyms: 2,3-dibromo-1,1,3,3-tetrafluoroprop-1-ene
Molecular Weight (g/mol): 271.84
Molecular Formula: C3Br2F4
InChi Key: HFEIAWIQHJLHCT-UHFFFAOYSA-N
SMILES: FC(F)=C(Br)C(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results