Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromo-1,1,3,3-tetrafluoropropene, TCI America™
SDP

Catalog No. D26715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26715G Supplier TCI America Supplier No. D26715G

Chemical Identifiers

CAS 666-40-0
Molecular Formula C3Br2F4
Molecular Weight (g/mol) 271.835
MDL Number MFCD00236663
InChI Key HFEIAWIQHJLHCT-UHFFFAOYSA-N
Synonym 2,3-dibromo-1,1,3,3-tetrafluoropropene, acmc-1azor, 2,3-dibromoperfluoropropene, perfluoro-2,3-dibromopropene, 1,1,3,3-tetrafluoro-2,3-dibromopropene, 2,3-dibromo-1,1,3,3-tetrafluoropropene-1
PubChem CID 2736793
IUPAC Name 2,3-dibromo-1,1,3,3-tetrafluoroprop-1-ene
SMILES C(=C(F)F)(C(F)(F)Br)Br

Specifications

Quantity 5 g
Formula Weight 271.83
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromo-1,1,3,3-tetrafluoropropene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.