accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66644-80-2

IUPAC Name: 2,3-dimethoxy-5-sulfamoylbenzoic acid
Synonyms: 2,3-dimethoxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol): 261.25
Molecular Formula: C9H11NO6S
InChi Key: YEVQOPOKMKTXMD-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(C(O)=O)=C1OC)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results