Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Aminosulfonyl)-2,3-dimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A21265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21265G 5 g
A212625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21265G Supplier TCI America Supplier No. A21265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66644-80-2
Molecular Formula C9H11NO6S
Molecular Weight (g/mol) 261.25
MDL Number MFCD01317561
InChI Key YEVQOPOKMKTXMD-UHFFFAOYSA-N
Synonym 2,3-Dimethoxy-5-sulfamoylbenzoic Acid
PubChem CID 3017698
IUPAC Name 2,3-dimethoxy-5-sulfamoylbenzoic acid
SMILES COC1=CC(=CC(C(O)=O)=C1OC)S(N)(=O)=O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 5 g
Formula Weight 261.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Aminosulfonyl)-2,3-dimethoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.