accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBrNHNOHO(E/Z)

CAS RN 667463-95-8

IUPAC Name: 6'-bromo-3-(hydroxyamino)-1'-methyl-1',2'-dihydro-[2,3'-biindolyliden]-2'-one
Synonyms: 6'-bromo-3-(hydroxyamino)-1'-methyl-[2,3'-biindolyliden]-2'-one
Molecular Weight (g/mol): 370.21
Molecular Formula: C17H12BrN3O2
InChi Key: WWVVQXIBSZPELF-UHFFFAOYSA-N
SMILES: CN1C(=O)C(C2=C1C=C(Br)C=C2)=C1N=C2C=CC=CC2=C1NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results