Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MeBIO, R&D Systems™
SDP

Catalog No. 387310 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-731-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 38-731-0 Supplier R&D Systems Supplier No. 3873/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inactive analog of BIO (Cat. No. 3194)

Control analog of 6-bromoindirubin-3'-oxime ( BIO ). Displays minimal activity against CDK1/Cyclin B, GSK-3 α/β, and CDK5/p25 (IC50 values are 92.0, 44-100 and >100 μM respectively). Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.

Active Analog also available.

Chemical Identifiers

CAS 667463-95-8
Molecular Formula C17H12BrN3O2
Molecular Weight (g/mol) 370.206
InChI Key WWVVQXIBSZPELF-JQIJEIRASA-N
Synonym mebio, gsk-3 inhibitor ix, control, mebio
PubChem CID 73952600
IUPAC Name (3E)-6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1-methylindol-2-one
SMILES CN1C2=C(C=CC(=C2)Br)C(=C3C(=C4C=CC=CC4=N3)NO)C1=O

Specifications

Quantity 10 mg
Formula Weight 370.2
Percent Purity >99%
Product Line Tocris
Chemical Name or Material MeBIO
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.