accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOONNN

CAS RN 66761-27-1

IUPAC Name: 3-azido-5-hydroxybenzoic acid
Synonyms: 3-azido-5-hydroxybenzoic acid
Molecular Weight (g/mol): 179.14
Molecular Formula: C7H5N3O3
InChi Key: IMKRSVNNWZYMKI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=CC(=C1)N=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results