accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHN(S)(R)NH2(R)OO(R)(S)NH2(R)NH2(S)O(R)O(R)H3CHO(R)HNCH3(R)OH(S)HOOHSOHOO

CAS RN 66803-19-8

IUPAC Name: (2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4,6-diamino-3-{[(2R,3R,6S)-3-amino-6-[(methylamino)methyl]oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol; sulfuric acid
Synonyms: micronomicin
Molecular Weight (g/mol): 561.65
Molecular Formula: C20H43N5O11S
InChi Key: MQDGQSCLOYLSEK-SCFBDNQUSA-N
SMILES: OS(O)(=O)=O.CNC[C@@H]1CC[C@@H](N)[C@@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)O1
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results