accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CCH3OOO

CAS RN 66872-74-0

IUPAC Name: sodium 3-methyl-2-oxopentanoate
Synonyms: sodium 3-methyl-2-oxopentanoate
Molecular Weight (g/mol): 152.13
Molecular Formula: C6H9NaO3
InChi Key: SMDJDLCNOXJGKC-UHFFFAOYNA-M
SMILES: [Na+].CCC(C)C(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results