accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNH3CHOOH

CAS RN 669072-93-9

IUPAC Name: (9-ethyl-9H-carbazol-3-yl)boronic acid
Synonyms: 9-ethylcarbazol-3-ylboronic acid
Molecular Weight (g/mol): 239.08
Molecular Formula: C14H14BNO2
InChi Key: JUBKLYCRFZTXBA-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results