CAS RN 6698-26-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 2',5'-Dideoxyadenosine
Cell-permeable, non-competitive adenylate cyclase inhibitor (IC50 = 3 μM), that binds to the adenosine P1 binding site.