CAS RN 67-17-5
CAS RN 67-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute, Ricca Chemical
Molecular Formula: C2H6O InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO