Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute, Ricca Chemical
SDP

Catalog No. 65901 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
3.8 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6590-1 4 L Natural Poly Bottle
6590-16 500 mL Natural Poly Bottle
6590-32 1 L Natural Poly Bottle
659025 10 L Cubitainer
R659000038 3.8 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6590-1 Supplier Ricca Chemical Company Supplier No. 65901

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Chemical Identifiers

Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS 67-17-5, 67-63-0, 67-56-1
CAS Min % 90.0, 4.0, 4.0
CAS Max % 91.0, 6.0, 5.0
Appearance Colorless Liquid
Quantity 4 L
Packaging Natural Poly Bottle
Residue after Evaporation max. 0.001%
Water max. 0.5 %
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 76°C
Molecular Weight (g/mol) 46.069
Percent Purity 89.5 to 91.5%
Grade ACS
Chemical Name or Material Alcohol, Reagent , Anhyd., Absolute, Denatured alcohol
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.