accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OBrCH3O(S)(S)(S)NH3CH

CAS RN 6700-34-1

IUPAC Name: hydrogen (1S,9S,10S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.0¹,¹⁰.0²,⁷]heptadeca-2,4,6-triene hydrate bromide
Synonyms: dextromethorphan hydrobromide
Molecular Weight (g/mol): 370.33
Molecular Formula: C18H28BrNO2
InChi Key: STTADZBLEUMJRG-IKNOHUQMSA-N
SMILES: [H+].O.[Br-].COC1=CC=C2C[C@H]3[C@H]4CCCC[C@@]4(CCN3C)C2=C1
Grade 
  • (1)
  • (12)
Molecular Weight (g/mol) 
  • (5)
  • (7)
Percent Purity 
  • (5)
  • (7)
Physical Form 
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (12)
  • (5)
  • (1)
  • (7)
  • (7)
  • (1)
  • (12)
  • (5)
  • (7)
  • (5)
  • (7)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)