CAS RN 67006-34-2
IUPAC Name:
2-[2-hydroxy-3-(2-hydroxyethanesulfonyl)propanesulfonyl]ethan-1-ol
Synonyms:
2-[2-hydroxy-3-(2-hydroxyethanesulfonyl)propanesulfonyl]ethanol
Molecular Weight (g/mol):
276.32
Molecular Formula:
C7H16O7S2
InChi Key:
CVFXDAXIZHJZAH-UHFFFAOYSA-N
SMILES:
OCCS(=O)(=O)CC(O)CS(=O)(=O)CCO
1
–
1
of
1
results
1
1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol 93.0+%, TCI America™
CAS: 67006-34-2 Molecular Formula: C7H16O7S2 Molecular Weight (g/mol): 276.318 InChI Key: CVFXDAXIZHJZAH-UHFFFAOYSA-N PubChem CID: 11437335 IUPAC Name: 1,3-bis(2-hydroxyethylsulfonyl)propan-2-ol SMILES: C(CS(=O)(=O)CC(CS(=O)(=O)CCO)O)O