accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 67071-94-7

IUPAC Name: heptacosa-10,12-diynoic acid
Synonyms: heptacosa-10,12-diynoic acid
Molecular Weight (g/mol): 402.66
Molecular Formula: C27H46O2
InChi Key: DWPBEQMBEZDLDV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results