accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6712-98-7

CH3OHNHOOH

CAS RN 6712-98-7

IUPAC Name: 1-[bis(2-hydroxyethyl)amino]propan-2-ol
Synonyms: 1-[bis(2-hydroxyethyl)amino]propan-2-ol
Molecular Weight (g/mol): 163.22
Molecular Formula: C7H17NO3
InChi Key: ZFECCYLNALETDE-UHFFFAOYNA-N
SMILES: CC(O)CN(CCO)CCO
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)