CAS RN 6724-53-4
IUPAC Name:
(2Z)-but-2-enedioic acid; 2-(2,2-dicyclohexylethyl)piperidine
Synonyms:
maleic acid; perhexiline
Molecular Weight (g/mol):
393.57
Molecular Formula:
C23H39NO4
InChi Key:
JDZOTSLZMQDFLG-BTJKTKAUNA-N
SMILES:
OC(=O)\C=C/C(O)=O.C(C(C1CCCCC1)C1CCCCC1)C1CCCCN1
1
–
1
of
1
results
1
R&D Systems™ Perhexiline maleate, >99%
CAS: 6724-53-4 Molecular Formula: C23H39NO4