accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNONH2

CAS RN 67242-60-8

IUPAC Name: 2-methyl-1H-indole-3-carboxamide
Synonyms: 2-methyl-1H-indole-3-carboxamide
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: QKWLFJZXUBMFLU-UHFFFAOYSA-N
SMILES: CC1=C(C(N)=O)C2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results