CAS RN 67287-39-2
IUPAC Name:
6-chloro-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Synonyms:
6-chloro-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrobromide
Molecular Weight (g/mol):
370.67
Molecular Formula:
C16H17BrClNO2
InChi Key:
RMIJGBMRNYUZRG-UHFFFAOYNA-N
SMILES:
Br.OC1=C(O)C(Cl)=C2CCNCC(C3=CC=CC=C3)C2=C1
1
–
1
of
1
results
1
R&D Systems™ SKF 81297 hydrobromide
CAS: 67287-39-2 Molecular Formula: C16H17BrClNO2