CAS RN 67469-75-4
IUPAC Name:
1-[2-(diphenylmethoxy)ethyl]-4-(3-phenylprop-2-en-1-yl)piperazine dihydrochloride
Synonyms:
1-[2-(diphenylmethoxy)ethyl]-4-(3-phenylprop-2-en-1-yl)piperazine dihydrochloride
Molecular Weight (g/mol):
485.49
Molecular Formula:
C28H34Cl2N2O
InChi Key:
HJDXBLLTRMCYNC-UHFFFAOYSA-N
SMILES:
Cl.Cl.C(CN1CCN(CC=CC2=CC=CC=C2)CC1)OC(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ GBR 12783 dihydrochloride
CAS: 67469-75-4 Molecular Formula: C28H34Cl2N2O