accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)(S)(R)H3CCH3OCH3HO

CAS RN 6754-20-7

IUPAC Name: (1R,4aS,8aS)-5,5,8a-trimethyl-1,4,4a,5,6,7,8,8a-octahydronaphthalene-1,2-dicarbaldehyde
Synonyms: polygodial tadeonal epipolygodial drim-7-ene-11,12-dial (-)-tadeonal poligodial (-)-polygodial
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: AZJUJOFIHHNCSV-KCQAQPDRSA-N
SMILES: CC1(C)CCC[C@@]2(C)[C@H]1CC=C(C=O)[C@@H]2C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results