accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6754-58-1

IUPAC Name: (2E)-1-[2,4-dihydroxy-6-methoxy-3-(3-methylbut-2-en-1-yl)phenyl]-3-(4-hydroxyphenyl)prop-2-en-1-one
Synonyms: xanthohumol
Molecular Weight (g/mol): 354.4
Molecular Formula: C21H22O5
InChi Key: ORXQGKIUCDPEAJ-YRNVUSSQSA-N
SMILES: COC1=CC(O)=C(CC=C(C)C)C(O)=C1C(=O)\C=C\C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results