Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xanthohumol 97.0+%, TCI America™
SDP

Catalog No. X006825MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X006825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X006825MG Supplier TCI America Supplier No. X006825MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6754-58-1
Molecular Formula C21H22O5
Molecular Weight (g/mol) 354.402
MDL Number MFCD00210576
InChI Key ORXQGKIUCDPEAJ-YRNVUSSQSA-N
Synonym xanthohumol, 1-2,4-dihydroxy-6-methoxy-3-3-methylbut-2-en-1-yl phenyl-3-4-hydroxyphenyl prop-2-en-1-one, unii-t4467yt1nt, 2',4,4'-trihydroxy-6'-methoxy-3'-prenylchalcone, 1-2,4-dihydroxy-6-methoxy-3-3-methyl-2-butenyl phenyl-3-4-hydroxyphenyl-2-propen-1-one, 2e-1-2,4-dihydroxy-6-methoxy-3-3-methylbut-2-en-1-yl phenyl-3-4-hydroxyphenyl prop-2-en-1-one, e-1-2,4-dihydroxy-6-methoxy-3-3-methyl-2-butenyl phenyl-3-4-hydroxyphenyl propenone, e-1-2,4-dihydroxy-6-methoxy-3-3-methylbut-2-enyl phenyl-3-4-hydroxyphenyl prop-2-en-1-one, xanthohumol from hop humulus lupulus, 2',4,4'-trihydroxy-3'-prenyl-6'-methoxychalcone
PubChem CID 639665
ChEBI CHEBI:66331
IUPAC Name (E)-1-[2,4-dihydroxy-6-methoxy-3-(3-methylbut-2-enyl)phenyl]-3-(4-hydroxyphenyl)prop-2-en-1-one
SMILES CC(=CCC1=C(C=C(C(=C1O)C(=O)C=CC2=CC=C(C=C2)O)OC)O)C

Specifications

Melting Point 159°C
Quantity 25 mg
Formula Weight 354.40
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Xanthohumol
RTECSNumber UD5574117
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.