CAS RN 675599-62-9
IUPAC Name:
N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]but-2-en-1-yl}-4-(pyridin-2-yl)benzamide dihydrochloride
Synonyms:
N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]but-2-en-1-yl}-4-(pyridin-2-yl)benzamide dihydrochloride
Molecular Weight (g/mol):
554.34
Molecular Formula:
C26H28Cl4N4O
InChi Key:
DACFFMAXBWGOKQ-UHFFFAOYSA-N
SMILES:
Cl.Cl.ClC1=CC=CC(N2CCN(CC=CCNC(=O)C3=CC=C(C=C3)C3=CC=CC=N3)CC2)=C1Cl
1
–
1
of
1
results
1
R&D Systems™ PG 01037 dihydrochloride
CAS: 675599-62-9 Molecular Formula: C26H28Cl4N4O