CAS RN 67567-26-4
CAS RN 67567-26-4
IUPAC Name:
4-bromo-2,6-difluoroaniline
Synonyms:
4-bromo-2,6-difluoroaniline
Molecular Weight (g/mol):
208.01
Molecular Formula:
C6H4BrF2N
InChi Key:
BFQSQUAVMNHOEF-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=C(Br)C=C1F
1
–
1
of
1
results
1
4-Bromo-2,6-difluoroaniline 98.0+%, TCI America™
CAS: 67567-26-4 Molecular Formula: C6H4BrF2N Molecular Weight (g/mol): 208.006 MDL Number: MFCD00013389 InChI Key: BFQSQUAVMNHOEF-UHFFFAOYSA-N Synonym: 2,6-difluoro-4-bromoaniline,4-bromo-2,6-difluorobenzenamine,benzenamine, 4-bromo-2,6-difluoro,4-bromo-2,6-difluoro-phenylamine,4-bromo-2,6-difluoro-aniline,4-bromo-2,6-difluorophenylamine,buttpark 22\07-66,4-bromo-2,6-difluorophenyl amine,pubchem2929,acmc-1begp PubChem CID: 610191 IUPAC Name: 4-bromo-2,6-difluoroaniline SMILES: C1=C(C=C(C(=C1F)N)F)Br