CAS RN 676-97-1
CAS RN 676-97-1
IUPAC Name:
methylphosphonoyl dichloride
Synonyms:
methylphosphonic acid dichloride
Molecular Weight (g/mol):
132.91
Molecular Formula:
CH3Cl2OP
InChi Key:
SCLFRABIDYGTAZ-UHFFFAOYSA-N
SMILES:
CP(Cl)(Cl)=O
1
–
2
of
2
results
1
Promotions Available
Sigma Aldrich Methylphosphonic Dichloride
Sigma Aldrich Methylphosphonic Dichloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.