accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 67634-00-8

IUPAC Name: prop-2-en-1-yl 2-(3-methylbutoxy)acetate
Synonyms: isoamyl allylglycolate
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: XCWPXUNHSPOFGV-UHFFFAOYSA-N
SMILES: CC(C)CCOCC(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results